Learn More
Epigentek Inc EpiQuik HDAC Activity/Inhibition Assay Kit (Colorimetric) (96 assays)
SDP
Catalog No. 501096337
Encompass_Preferred
The EpiQuik HDAC Activity/Inhibition Assay Kit is a ready-to-use set of essential components needed to measure HDAC activity/inhibition safely and quickly. It is suitable for measuring HDAC activity/inhibition from a broad range of species including mammalian cells/tissues, plants, and bacteria.
Catalog No. 50-109-6337 Supplier Epigentek Inc Supplier No. P400296
The EpiQuik HDAC Activity/Inhibition Assay Kit is a ready-to-use set of essential components needed to measure HDAC activity/inhibition safely and quickly. It is suitable for measuring HDAC activity/inhibition from a broad range of species including mammalian cells/tissues, plants, and bacteria.