Learn More
Epigentek Inc EpiQuik Histone H3 Citrullination ELISA Kit (Colorimetric) (96 assays)
SDP
Supplier:  Epigentek Inc P309596
Encompass_Preferred
For ELISA-like measurement of total citrullinated histone H3 (at arginines R2, R8, R17)
Catalog No. 50-125-0834