Learn More
Epigentek Inc EpiQuik Histone H3 Modification Multiplex Assay Kit (Colorimetric) (96 assays)
SDP
Catalog No. NC0894762
Encompass_Preferred
For ELISA-like quantitation of 21 different histone H3 modification patterns on the same plate
Catalog No. NC0894762 Supplier Epigentek Inc Supplier No. P310096
For ELISA-like quantitation of 21 different histone H3 modification patterns on the same plate