Learn More
Epigentek Inc EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3K4) (48 assays)
SDP
Catalog No. 501096178
Encompass_Preferred
The EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K4) is a convenient set of tools that allows the experimenter to measure the activity or inhibition of individual histone methyltransferase that specifically target histone H3 at lysine 4 (H3-K4). The kit is ready-to-use and provides all the essential components needed to carry out a successful HMT activity/inhibition experiment without the need for radioactivity or any special equipment.
Catalog No. 50-109-6178 Supplier Epigentek Inc Supplier No. P30021
The EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K4) is a convenient set of tools that allows the experimenter to measure the activity or inhibition of individual histone methyltransferase that specifically target histone H3 at lysine 4 (H3-K4). The kit is ready-to-use and provides all the essential components needed to carry out a successful HMT activity/inhibition experiment without the need for radioactivity or any special equipment.