Learn More
Epigentek Inc EpiQuik m6A RNA Methylation Quantification Kit (Colorimetric) (96 reactions)
SDP
Catalog No. NC0620304
Encompass_Preferred
For absorbance-based quantitation of N6-methyladenosine in RNA in an ELISA-like format
Catalog No. NC0620304 Supplier Epigentek Inc Supplier No. P900596
Add to Cart
Add to Cart
For absorbance-based quantitation of N6-methyladenosine in RNA in an ELISA-like format