Learn More
Epigentek Inc EpiQuik Total Histone Extraction Kit (100 extractions)
SDP
Catalog No. NC9892093
Encompass_Preferred
For isolating histone proteins with post-translational modifications (PTMs) intact
Catalog No. NC9892093 Supplier Epigentek Inc Supplier No. OP0006100
For isolating histone proteins with post-translational modifications (PTMs) intact