Learn More
Selleck Chemical LLC Epirubicin HCl S1223-10mM/1mL
SDP
Catalog No. 503108267
Encompass_Preferred
Epirubicin HCl a semisynthetic L-arabino derivative of doxorubicin is an antineoplastic agent by inhibiting Topoisomerase Epirubicin induces apoptosis
Catalog No. 50-310-8267 Supplier Selleck Chemical LLC Supplier No. S122310MM/1ML
Add to Cart
Add to Cart
Epirubicin HCl a semisynthetic L-arabino derivative of doxorubicin is an antineoplastic agent by inhibiting Topoisomerase Epirubicin induces apoptosis