Learn More
Selleck Chemical LLC Epirubicin HCl 5mg 56390-09-1 4'-epidoxorubicin HCl
SDP
Catalog No. 501361043
Encompass_Preferred
Epirubicin HCl (4'-epidoxorubicin), a semisynthetic L-arabino derivative of doxorubicin, is an antineoplastic agent by inhibiting Topoisomerase. Epirubicin induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1043 Supplier Selleck Chemical LLC Supplier No. S12235MG
Epirubicin HCl (4'-epidoxorubicin), a semisynthetic L-arabino derivative of doxorubicin, is an antineoplastic agent by inhibiting Topoisomerase. Epirubicin induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.