Learn More
Apexbio Technology LLC Epirubicin HCl, 5mg. Cas: 56390-09-1 MFCD: MFCD00941448. Antibiotic antitumor agent
SDP
Supplier:  Apexbio Technology LLC A24515
Encompass_Preferred
Epirubicin is an inhibitor of DNA topoisomerase (TOPII) [1].Epirubicin belongs to the Anthracylines chemical class, it is a sort of DNA topoisomerase poison. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4743