Learn More
Selleck Chemical LLC Epirubicin HCl 25mg 56390-09-1 4'-epidoxorubicin HCl
SDP
Catalog No. 507110
Encompass_Preferred
Epirubicin HCl (4'-epidoxorubicin), a semisynthetic L-arabino derivative of doxorubicin, is an antineoplastic agent by inhibiting Topoisomerase. Epirubicin induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-711-0 Supplier Selleck Chemical LLC Supplier No. S122325MG
Epirubicin HCl (4'-epidoxorubicin), a semisynthetic L-arabino derivative of doxorubicin, is an antineoplastic agent by inhibiting Topoisomerase. Epirubicin induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.