Learn More
Medchemexpress LLC Epirubicin hydrochloride | 56390-09-1 | >99.2% | 579.98 | 100 MG
SDP
Catalog No. 5000350953
Encompass_Preferred
Epirubicin hydrochloride is a semisynthetic L-arabino derivative of doxorubicin, functioning as an antineoplastic agent by inhibiting Topoisomerase and DNA/RNA synthesis. It also acts as a Forkhead box protein p3 (Foxp3) inhibitor, suppressing regulatory T cell activity. Its antitumor effects involve forming a complex with DNA, causing damage and interfering with essential synthesis processes.
  • Acts as an antineoplastic agent.
  • Inhibits Topoisomerase, DNA, and RNA synthesis.
  • Functions as a Forkhead box protein p3 (Foxp3) inhibitor.
  • Suppresses regulatory T cell activity.
  • Causes DNA damage by complexing with DNA.
  • Cytotoxic to various cancer cells, including Hepatoma G2 cells (IC50 1.6 μg/mL).
  • Induces apoptosis and alters antioxidant enzyme activity in Hep G2 cells.
  • Clinically active against a wide range of tumor types.
Catalog No. 50-003-50953 Supplier Medchemexpress LLC Supplier No. HY13624A100MG
Add to Cart
Add to Cart
Epirubicin hydrochloride is a semisynthetic L-arabino derivative of doxorubicin, functioning as an antineoplastic agent by inhibiting Topoisomerase and DNA/RNA synthesis. It also acts as a Forkhead box protein p3 (Foxp3) inhibitor, suppressing regulatory T cell activity. Its antitumor effects involve forming a complex with DNA, causing damage and interfering with essential synthesis processes.
  • Acts as an antineoplastic agent.
  • Inhibits Topoisomerase, DNA, and RNA synthesis.
  • Functions as a Forkhead box protein p3 (Foxp3) inhibitor.
  • Suppresses regulatory T cell activity.
  • Causes DNA damage by complexing with DNA.
  • Cytotoxic to various cancer cells, including Hepatoma G2 cells (IC50 1.6 μg/mL).
  • Induces apoptosis and alters antioxidant enzyme activity in Hep G2 cells.
  • Clinically active against a wide range of tumor types.