Learn More
Medchemexpress LLC Epirubicin hydrochloride | 56390-09-1 | 99.2% | 579.98 | 25 MG
SDP
Catalog No. 5000352308
Encompass_Preferred
Epirubicin hydrochloride (4'-Epidoxorubicin hydrochloride) is a semisynthetic L-arabino derivative of doxorubicin. It acts as an antineoplastic agent by inhibiting Topoisomerase, and also inhibits DNA and RNA synthesis. Furthermore, it functions as a Forkhead box protein p3 (Foxp3) inhibitor, leading to the inhibition of regulatory T cell activity. This product is an analytical standard intended for research and analytical applications.
  • Semisynthetic L-arabino derivative of doxorubicin
  • Inhibits Topoisomerase
  • Inhibits DNA and RNA synthesis
  • Functions as a Forkhead box protein p3 (Foxp3) inhibitor
  • Inhibits regulatory T cell activity
  • Analytical standard for research and analytical applications
Catalog No. 50-003-52308 Supplier Medchemexpress LLC Supplier No. 25MGHY13624A
Epirubicin hydrochloride (4'-Epidoxorubicin hydrochloride) is a semisynthetic L-arabino derivative of doxorubicin. It acts as an antineoplastic agent by inhibiting Topoisomerase, and also inhibits DNA and RNA synthesis. Furthermore, it functions as a Forkhead box protein p3 (Foxp3) inhibitor, leading to the inhibition of regulatory T cell activity. This product is an analytical standard intended for research and analytical applications.
  • Semisynthetic L-arabino derivative of doxorubicin
  • Inhibits Topoisomerase
  • Inhibits DNA and RNA synthesis
  • Functions as a Forkhead box protein p3 (Foxp3) inhibitor
  • Inhibits regulatory T cell activity
  • Analytical standard for research and analytical applications