Learn More
Medchemexpress LLC Epirubicin hydrochloride (Standard) | 56390-09-1 | 99.9% | 579.98 | 25 MG
SDP
Catalog No. 5000352309
Encompass_Preferred
Epirubicin hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It is a semisynthetic L-arabino derivative of doxorubicin, acting as an antineoplastic agent. This compound inhibits Topoisomerase, DNA, and RNA synthesis, and also functions as a Forkhead box protein p3 (Foxp3) inhibitor.
  • Analytical standard for research and analytical applications
  • Semisynthetic L-arabino derivative of doxorubicin
  • Antineoplastic agent
  • Inhibits Topoisomerase
  • Inhibits DNA and RNA synthesis
  • Foxp3 inhibitor, inhibiting regulatory T cell activity
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-52309 Supplier Medchemexpress LLC Supplier No. 25MGHY13624AR
Epirubicin hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It is a semisynthetic L-arabino derivative of doxorubicin, acting as an antineoplastic agent. This compound inhibits Topoisomerase, DNA, and RNA synthesis, and also functions as a Forkhead box protein p3 (Foxp3) inhibitor.
  • Analytical standard for research and analytical applications
  • Semisynthetic L-arabino derivative of doxorubicin
  • Antineoplastic agent
  • Inhibits Topoisomerase
  • Inhibits DNA and RNA synthesis
  • Foxp3 inhibitor, inhibiting regulatory T cell activity
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS