Learn More
Medchemexpress LLC Epirubicin hydrochloride (standard) | 56390-09-1 | 99.91% | 579.98 | 100 MG
SDP
Catalog No. 5000349745
Encompass_Preferred
Epirubicin hydrochloride (Standard) is an analytical standard and a semisynthetic L-arabino derivative of doxorubicin, primarily used in research and analytical applications. It functions as an antineoplastic agent by inhibiting Topoisomerase and also inhibits DNA and RNA synthesis. Additionally, it acts as a Forkhead box protein p3 (Foxp3) inhibitor, effectively inhibiting regulatory T cell activity.
  • Analytical standard for research and analytical applications
  • Semisynthetic L-arabino derivative of doxorubicin
  • Acts as an antineoplastic agent
  • Inhibits Topoisomerase, DNA, and RNA synthesis
  • Functions as a Forkhead box protein p3 (Foxp3) inhibitor
  • Inhibits regulatory T cell activity
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)
Catalog No. 50-003-49745 Supplier Medchemexpress LLC Supplier No. HY13624AR100MG
Epirubicin hydrochloride (Standard) is an analytical standard and a semisynthetic L-arabino derivative of doxorubicin, primarily used in research and analytical applications. It functions as an antineoplastic agent by inhibiting Topoisomerase and also inhibits DNA and RNA synthesis. Additionally, it acts as a Forkhead box protein p3 (Foxp3) inhibitor, effectively inhibiting regulatory T cell activity.
  • Analytical standard for research and analytical applications
  • Semisynthetic L-arabino derivative of doxorubicin
  • Acts as an antineoplastic agent
  • Inhibits Topoisomerase, DNA, and RNA synthesis
  • Functions as a Forkhead box protein p3 (Foxp3) inhibitor
  • Inhibits regulatory T cell activity
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)