Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epirubicin hydrochloride, Tocris Bioscience™

Catalog No. 326010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-601-0 Supplier R&D Systems Supplier No. 3260/10
This item is not returnable. View return policy

Inhibits DNA synthesis and function. Inhibits DNA topoisomerase II

Antibiotic antitumor agent. Inhibits the synthesis and function of DNA (IC50 = 62.7 μM in rat glioblastoma cell lines) and inhibits the relaxing property of topoisomerase II.

Chemical Identifiers

CAS 56390-09-1
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.983
InChI Key MWWSFMDVAYGXBV-DPZLWMTJSA-N
Synonym (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-α-L-arabino-hexopyranosyl)oxy]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,12-naphthacenedione hydrochloride
PubChem CID 123134690
IUPAC Name (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl

Specifications

Quantity 10 mg
Formula Weight 579.98
Percent Purity >97%
Chemical Name or Material Epirubicin hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.