Learn More
Cayman Chemical Epirubicin (hydrochloride), Purity: ≥98%, CAS: 56390-09-1, 100 mg
SDP
Catalog No. 502955465
Encompass_Preferred
A stereoisomer of the antitumor anthracycline doxorubicin that undergoes β-glucuronidation, which partially detoxifies the dose-limiting side effects that are present with doxorubicin; equally cytotoxic to HeLa cells compared to doxorubicin (ID50s = 9 μM)
Catalog No. 50-295-5465 Supplier Cayman Chemical Supplier No. 12091100 MG
A stereoisomer of the antitumor anthracycline doxorubicin that undergoes β-glucuronidation, which partially detoxifies the dose-limiting side effects that are present with doxorubicin; equally cytotoxic to HeLa cells compared to doxorubicin (ID50s = 9 μM)