Learn More
Cayman Chemical EpIrubIcInhydrochlorIde 10mg
SDP
Catalog No. 502954925
Encompass_Preferred
A stereoisomer of the antitumor anthracycline doxorubicin that undergoes β-glucuronidation, which partially detoxifies the dose-limiting side effects that are present with doxorubicin; equally cytotoxic to HeLa cells compared to doxorubicin (ID50s = 9 μM)
Catalog No. 50-295-4925 Supplier Cayman Chemical Supplier No. 1209110 MG
A stereoisomer of the antitumor anthracycline doxorubicin that undergoes β-glucuronidation, which partially detoxifies the dose-limiting side effects that are present with doxorubicin; equally cytotoxic to HeLa cells compared to doxorubicin (ID50s = 9 μM)