Learn More
LGC GENOMICS LLC (Lucigen) EpiScript RNase H- Reverse Transcriptase, 25,000 units @ 200 U/LAn optimised reverse transcriptase system for the production of full-length cDNA.
Catalog No. NC1181385
Encompass
EpiScript™ Reverse Transcriptase an alternative to SuperScript II Reverse Transcriptase, is a recombinant MMLV reverse transcriptase with reduced RNase H activityProduce full-length cDNA with RNAse H-mutantAchieve higher specificity at elevated temperatures, up to 55 CKit Contents: EpiScript RNase H- Reverse Transcriptase, 10X Reaction Buffer, 100 mM DTT.
This item is not returnable. View return policy
Catalog No. NC1181385 Supplier LGC GENOMICS LLC Supplier No. ERT12925K
EpiScript™ Reverse Transcriptase an alternative to SuperScript II Reverse Transcriptase, is a recombinant MMLV reverse transcriptase with reduced RNase H activityProduce full-length cDNA with RNAse H-mutantAchieve higher specificity at elevated temperatures, up to 55 CKit Contents: EpiScript RNase H- Reverse Transcriptase, 10X Reaction Buffer, 100 mM DTT.