Learn More
LGC GENOMICS LLC (Lucigen) EpiScript™ RNase H- Reverse Transcriptase Kit, 200 U/L, 10,000 Units, 50 LAn optimised reverse transcriptase system for the production of full-length cDNA.
Catalog No. NC2160305
Encompass
EpiScript™ Reverse Transcriptase an alternative to SuperScript II Reverse Transcriptase, is a recombinant MMLV reverse transcriptase with reduced RNase H activityProduce full-length cDNA with RNAse H-mutantAchieve higher specificity at elevated temperatures, up to 55 CKit Contents: EpiScript RNase H- Reverse Transcriptase, 10X Reaction Buffer, 100 mM DTT.
This item is not returnable. View return policy
Catalog No. NC2160305 Supplier LGC GENOMICS LLC Supplier No. ERT12910K
EpiScript™ Reverse Transcriptase an alternative to SuperScript II Reverse Transcriptase, is a recombinant MMLV reverse transcriptase with reduced RNase H activityProduce full-length cDNA with RNAse H-mutantAchieve higher specificity at elevated temperatures, up to 55 CKit Contents: EpiScript RNase H- Reverse Transcriptase, 10X Reaction Buffer, 100 mM DTT.