Learn More
Medchemexpress LLC Episilvestrol | 697235-39-5 | MFCD25541744 | C34H38O13 | 1 MG
SDP
Catalog No. 501873204
Encompass_Preferred
Episilvestrol is a derivative of silvestrol, isolated from the fruits and twigs of Aglaia perviridis. It functions as a specific eIF4A-targeting translation inhibitor, demonstrating significant antitumor activity.
  • Specific eIF4A-targeting translation inhibitor
  • Exhibits antitumor activity
  • Cytotoxic against human cancer cell lines including Lu1, LNCaP, MCF-7, and HUVEC
  • Suppresses HK1 cells and EBV-positive C666.1 NPC cells
  • Soluble in DMSO
Catalog No. 50-187-3204 Supplier Medchemexpress LLC Supplier No. HY153591MG
Episilvestrol is a derivative of silvestrol, isolated from the fruits and twigs of Aglaia perviridis. It functions as a specific eIF4A-targeting translation inhibitor, demonstrating significant antitumor activity.
  • Specific eIF4A-targeting translation inhibitor
  • Exhibits antitumor activity
  • Cytotoxic against human cancer cell lines including Lu1, LNCaP, MCF-7, and HUVEC
  • Suppresses HK1 cells and EBV-positive C666.1 NPC cells
  • Soluble in DMSO