Learn More
Sciencell Research Laboratories EPITHELEAL CELL MEDIUM
Catalog No. NC9657977
Encompass
Epithelial Cell Medium is a complete medium designed for optimal growth of normal human epithelial cells in vitro. It is shipped as a kit with epithelial cell growth supplement and Pen/Strep solution.
Catalog No. NC9657977 Supplier Sciencell Research Laboratories Supplier No. 4101
Add to Cart
Add to Cart
Epithelial Cell Medium is a complete medium designed for optimal growth of normal human epithelial cells in vitro. It is shipped as a kit with epithelial cell growth supplement and Pen/Strep solution.