Learn More
Sciencell Research Laboratories EPITHELIAL CELL MEDM GLUC FREE
Catalog No. NC2221675
Encompass
Epithelial Cell Medium Glucose free media.EpiCM consists of 500 ml of basal medium, 10 ml of fetal bovine serum (FBS, Cat. No. 0010), 5 ml of epithelial cell growth supplement (EpiCGS, Cat. No. 4152), and 5 ml of Antibiotic Solution (P/S, Cat. No. 0503).
This item is not returnable. View return policy
Catalog No. NC2221675 Supplier Sciencell Research Laboratories Supplier No. 4101GF
Epithelial Cell Medium Glucose free media.EpiCM consists of 500 ml of basal medium, 10 ml of fetal bovine serum (FBS, Cat. No. 0010), 5 ml of epithelial cell growth supplement (EpiCGS, Cat. No. 4152), and 5 ml of Antibiotic Solution (P/S, Cat. No. 0503).