Learn More
Biogenex Laboratories Epithelial : MOC-31 : 6mL
SDP
Supplier:  Biogenex Laboratories AM3165M
Encompass_Preferred
Monoclonal antibody MOC-31 recognizes the cluster 2 antigen which is a40 kD transmembrane glycoprotein present on the membrane of epithelialcells. It is a highly reliable and sensitive marker to distinguish betweenreactive epithelial and mesothelial cells. Since MOC-31 reacts withvirtually all normal epithelia and adenocarcinomas but not withmesothelial cells, this antibody can serve as a reliable marker fordetermining the origin of pleural and peritoneal tumors.
Catalog No. 50-197-8835