Learn More
Medchemexpress LLC Epitinib succinate | 2252334-12-4 | 99.0% | 548.59 | 1 ML
SDP
Catalog No. 5000352009
Encompass_Preferred
Epitinib succinate is an orally active and selective epidermal growth factor receptor tyrosine kinase inhibitor (EGFR-TKI) designed for optimal brain penetration. It can be used for cancer research. Epitinib succinate is also a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing azide groups.
  • Orally active and selective epidermal growth factor receptor tyrosine kinase inhibitor
  • Designed for optimal brain penetration
  • Used for cancer research
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition
Catalog No. 50-003-52009 Supplier Medchemexpress LLC Supplier No. HY139300A10MM/1ML
Epitinib succinate is an orally active and selective epidermal growth factor receptor tyrosine kinase inhibitor (EGFR-TKI) designed for optimal brain penetration. It can be used for cancer research. Epitinib succinate is also a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing azide groups.
  • Orally active and selective epidermal growth factor receptor tyrosine kinase inhibitor
  • Designed for optimal brain penetration
  • Used for cancer research
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition