Learn More
Medchemexpress LLC Epitinib succinate | 2252334-12-4 | 99.0% | 548.59 | 25 MG
SDP
Catalog No. 5000349096
Encompass_Preferred
Epitinib succinate is an orally active and selective epidermal growth factor receptor tyrosine kinase inhibitor (EGFR-TKI) designed for optimal brain penetration. It can be used for cancer research.
  • Selective epidermal growth factor receptor tyrosine kinase inhibitor (EGFR-TKI)
  • Designed for optimal brain penetration
  • Can be used for cancer research
  • Click chemistry reagent with an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide groups
Catalog No. 50-003-49096 Supplier Medchemexpress LLC Supplier No. 25MGHY139300A
Epitinib succinate is an orally active and selective epidermal growth factor receptor tyrosine kinase inhibitor (EGFR-TKI) designed for optimal brain penetration. It can be used for cancer research.
  • Selective epidermal growth factor receptor tyrosine kinase inhibitor (EGFR-TKI)
  • Designed for optimal brain penetration
  • Can be used for cancer research
  • Click chemistry reagent with an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide groups