Learn More
TARGETMOL CHEMICALS INC EPITINIB SUCCINATE 5MG
SDP
Catalog No. 502580203
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Epitinib succinate (HMPL-813 succinate) is an orally active and brain penetration EGFR tyrosine kinase inhibitor and can be used in studies about cancer. purity: 99%
Catalog No. 50-258-0203 Supplier TARGETMOL CHEMICALS INC Supplier No. T359145MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Epitinib succinate (HMPL-813 succinate) is an orally active and brain penetration EGFR tyrosine kinase inhibitor and can be used in studies about cancer. purity: 99%