Learn More
Leica Biosystems Epitope Ret Soln pH8 x10 1L
Supplier:  Leica Biosystems RE7116CE
Encompass
Novocastra Epitope Retrieval Solutions are intended for Heat Induced Epitope Retrieval (HIER) on formalin-fixed, paraffin-embedded tissue sections as part of an immunohistochemical procedure. HIER using an appropriate pH solution improves the staining of some antibodies by exposing epitopes within tissue that have been masked during fixation. A universal HIER technique suitable for all epitopes does not exist. A combination of different heating methods and epitope retrieval solutions may be used to optimize unmasking of antigens where this technique is recommended. HIER is not recommended for all antibodies. Optimum conditions for epitope retrieval should be validated by the user, as these are dependent upon tissue, fixation and/or primary antibody. RE7119 is supplied as a 1 L volume, sufficient to prepare 10 L of working solution. It is a Tris/EDTA-based buffer and should be diluted with de-ionized water to prepare a working solution. For in vitro diagnostic use.
Catalog No. 50-255-1989