Learn More
Bioworld Epitope Unmasking Buffer, 10X, 250 mL
Catalog No. 50199077
Encompass_Preferred
A citric acid based solution designed to restore epitope structure and enhance antigen-antibody recognition in immunohistochemistry and immunofluorescence. It comes in a 10X concentrated form.
Catalog No. 50-199-077 Supplier Bioworld Supplier No. 217600051
A citric acid based solution designed to restore epitope structure and enhance antigen-antibody recognition in immunohistochemistry and immunofluorescence. It comes in a 10X concentrated form.