Learn More
Cayman Chemical EPI-X4 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502915989
Encompass_Preferred
An endogenous peptide CXCR4 antagonist; inhibits CXCL12-induced calcium mobilization and receptor internalization in HEK293 cells expressing CXCR4 from 0.8-1,000 µM; inhibits CXCL12-induced migration of Jurkat T cells and human CD34+ hematopoietic stem cells; reduces Cxcr4-dependent inflammatory cell airway infiltration in a mouse model of acute allergic airway hypereosinophilia; urinary levels are increased and inversely correlated with glomerular filtration rate (GFR) in patients with chronic kidney disease
Catalog No. 50-291-5989 Supplier Cayman Chemical Supplier No. 3791210 MG
An endogenous peptide CXCR4 antagonist; inhibits CXCL12-induced calcium mobilization and receptor internalization in HEK293 cells expressing CXCR4 from 0.8-1,000 µM; inhibits CXCL12-induced migration of Jurkat T cells and human CD34+ hematopoietic stem cells; reduces Cxcr4-dependent inflammatory cell airway infiltration in a mouse model of acute allergic airway hypereosinophilia; urinary levels are increased and inversely correlated with glomerular filtration rate (GFR) in patients with chronic kidney disease