Learn More
Cayman Chemical Eplerenon-d3 2.5mg
SDP
Supplier:  Cayman Chemical 316102.5 MG
Encompass_Preferred
An internal standard for the quantification of eplerenone by GC- or LC-MS
Catalog No. 50-299-1264