Learn More
Medchemexpress LLC HY-B0251 10mg Medchemexpress, Eplerenone CAS:107724-20-9 Purity:>98%
SDP
Catalog No. 502082425
Encompass_Preferred
Medchemexpress, HY-B0251 10mg Eplerenone CAS:107724-20-9 Eplerenone (Epoxymexrenone) is a selective, competitive and oreally active aldosterone antagonist with an IC50 of 138 nM. Eplerenone has low affinity for progesterone, androgen, estrogen and glucocorticoid receptors. Eplerenone can be used for hypertension and heart failure after myocardial infarction reserch. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2425 Supplier Medchemexpress LLC Supplier No. HYB025110MG
Medchemexpress, HY-B0251 10mg Eplerenone CAS:107724-20-9 Eplerenone (Epoxymexrenone) is a selective, competitive and oreally active aldosterone antagonist with an IC50 of 138 nM. Eplerenone has low affinity for progesterone, androgen, estrogen and glucocorticoid receptors. Eplerenone can be used for hypertension and heart failure after myocardial infarction reserch. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.