Learn More
Maine Molecular Quality Controls Inc eplex BCID-FP Negative
SDP
Catalog No. NC3886265
Encompass
The eplex BCID-FP Negative is intended for use as an external negative quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Candida albicans, Candida auris, Candida dubliniensis, Candida famata, Candida glabrata, Candida guilliermondii, Candida kefyr, Candida krusei, Candida lusitaniae, Candida parapsilosis, Candida tropicalis, Cryptococcus gattii, Cryptococcus neoformans, Fusarium, and Rhodotorula on the cobas eplex blood culture identification fungal pathogen (BCID-FP) panel performed on the cobas eplex system (Roche Diagnostics). eplex BCID-FP Negative contains no DNA.
Catalog No. NC3886265 Supplier Maine Molecular Quality Controls Inc Supplier No. M501376
The eplex BCID-FP Negative is intended for use as an external negative quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Candida albicans, Candida auris, Candida dubliniensis, Candida famata, Candida glabrata, Candida guilliermondii, Candida kefyr, Candida krusei, Candida lusitaniae, Candida parapsilosis, Candida tropicalis, Cryptococcus gattii, Cryptococcus neoformans, Fusarium, and Rhodotorula on the cobas eplex blood culture identification fungal pathogen (BCID-FP) panel performed on the cobas eplex system (Roche Diagnostics). eplex BCID-FP Negative contains no DNA.