Learn More
Maine Molecular Quality Controls Inc eplex RP2 Control M451
SDP
Supplier:  Maine Molecular Quality Controls Inc M251
Encompass
The FilmArray Ebola Control Panel M251 is intended for in vitro use as a quality control to monitor the presumptive detection of Ebola Zaire virus in whole blood specimens when tested by the FilmArray BioThreat-E test on the FilmArray instrument. The FilmArray BioThreat-E test is performed on the FilmArray instrument to detect RNA from the Ebola Zaire virus in specimens from individuals with signs and symptoms of Ebola virus infection in conjunction with epidemiological risk factors. This product does not contain any biological material of human origin.
Catalog No. NC3886264