Learn More
Medchemexpress LLC Eplivanserin hemifumarate | 130580-02-8 | 98.1% | 386.43 g·mol⁻1 | C19H21FN2O2·1/2C4H4O4 | 5 MG
SDP
Catalog No. 5000263309
Encompass_Preferred
Eplivanserin hemifumarate is a research-grade small molecule described as a potent, selective, orally active 5-HT2A receptor antagonist. Supplied as the hemifumarate salt in solid form, it is intended for preclinical and in vitro pharmacology studies and comes with an accompanying purity specification and solubility information for assay preparation.
  • Potent, selective 5-HT2A receptor antagonist suitable for receptor pharmacology studies.
  • Supplied as hemifumarate salt in solid form for handling and storage.
  • High purity (~98.1%) appropriate for research applications.
  • Soluble in DMSO at 25 mg/mL; sonication and gentle heating aid dissolution.
  • Available in small pack sizes tailored for preclinical and in vitro work.
Catalog No. 50-002-63309 Supplier Medchemexpress LLC Supplier No. HY1101295MG
Eplivanserin hemifumarate is a research-grade small molecule described as a potent, selective, orally active 5-HT2A receptor antagonist. Supplied as the hemifumarate salt in solid form, it is intended for preclinical and in vitro pharmacology studies and comes with an accompanying purity specification and solubility information for assay preparation.
  • Potent, selective 5-HT2A receptor antagonist suitable for receptor pharmacology studies.
  • Supplied as hemifumarate salt in solid form for handling and storage.
  • High purity (~98.1%) appropriate for research applications.
  • Soluble in DMSO at 25 mg/mL; sonication and gentle heating aid dissolution.
  • Available in small pack sizes tailored for preclinical and in vitro work.