Learn More
Medchemexpress LLC HY-10792A 10mg Medchemexpress, Eplivanserin (mixture) CAS:130581-13-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10792A10MG
Encompass_Preferred
Medchemexpress, HY-10792A 10mg Eplivanserin (mixture) CAS:130581-13-4 Eplivanserin mixture is a selective serotonin reuptake inhibitor and a 5-HT2A receptor antagonist, extracted from patent WO 2005/002578 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8483