Learn More
Cayman Chemical Epimedin C, Purity: ≥98%, CAS: 110642-44-9, 10 mg
SDP
Catalog No. 502946325
Encompass_Preferred
A flavonoid glycoside; scavenges DPPH radicals in a cell-free assay (IC50 = 142 µM); increases proliferation of UMR-106 osteoblasts by 30.4 and 41.6% at 0.1 and 10 µM, respectively; inhibits TPA-induced ear edema in mice (ED50 = 192 nmol/ear)
Catalog No. 50-294-6325 Supplier Cayman Chemical Supplier No. 3437510 MG
A flavonoid glycoside; scavenges DPPH radicals in a cell-free assay (IC50 = 142 µM); increases proliferation of UMR-106 osteoblasts by 30.4 and 41.6% at 0.1 and 10 µM, respectively; inhibits TPA-induced ear edema in mice (ED50 = 192 nmol/ear)