Learn More
MILLER STEPHENSON CHEMICAL COMPANY INC EPON Resin 862, 5 gal
SDP
Catalog No. NC1794985
Encompass_Preferred
Low viscosity, liquid epoxy resin manufactured from epichlorohydrin and Bisphenol-F. This resin contains no diluents or modifiers. EPON Resin 862 may be used as the sole epoxy resin or combined with other resins such as EPON Resin 828.
Catalog No. NC1794985 Supplier MILLER STEPHENSON CHEMICAL COMPANY INC Supplier No. EPON 862 5GAL
Low viscosity, liquid epoxy resin manufactured from epichlorohydrin and Bisphenol-F. This resin contains no diluents or modifiers. EPON Resin 862 may be used as the sole epoxy resin or combined with other resins such as EPON Resin 828.