Learn More
Enzo Life Sciences Erythropoietin alpha (human), (recombinant) (500U)
SDP
Supplier:  Enzo Life Sciences ADI908050500
Encompass_Preferred
Alternative name: EPO. Purity: ≥98% (SDS-PAGE; RP-HPLC). MW: Predicted MW of ~21kDa (single chain) and ~30kDa (glycosylated). Formulation: Lyophilized. Each milligram contains 5.8mg sodium chloride and 0.06mg citric acid. Reconstitute with sterile 18W-cm water not <100μg/mL. Source: Produced in CHO cells. Long Term Storage: -20°C. Applications: ELISA, WB in vitro Assay.
Catalog No. 50-200-7087