Learn More
Aviva Systems Biology Corp EPO Antibody - middle region (ARP44337_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44337_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPO. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPO. Alternate names: EP, MGC138142, MVCD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-62996