Learn More
STEMCELL Technologies EPO Antibody, Clone 16F1H11, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 01300
Encompass
Erythropoietin (EPO) is the major regulator of red blood cell production and is produced in the kidney in response to hypoxia. This antibody can be used for immunoassay and immunoaffinity isolation of human EPO and for neutralizing human, mouse and baboon EPO.
Catalog No. 50-197-4246