Learn More
Electron Microscopy Sciences Epo-Fix Embedding Kit Small Clear
SDP
Catalog No. 50980282
Encompass_Preferred
A two component epoxy resin, characterized by negligible shrinkage during curing and good mechanical properties in its hardened state. It is particularly suited for embedding hard samples of complicated shape and for vacuum impregnation. Samples such as paper, silicon, ceramics, metals, wafers, and chips are suitable to be used with Epo-Fix. Kit consists of: 1 kg resin, 120 g hardener, 15 cups, 15 stirrers, 1 x 5 mL and 20 mL syringes.
Catalog No. 50-980-282 Supplier Electron Microscopy Sciences Supplier No. 1232
A two component epoxy resin, characterized by negligible shrinkage during curing and good mechanical properties in its hardened state. It is particularly suited for embedding hard samples of complicated shape and for vacuum impregnation. Samples such as paper, silicon, ceramics, metals, wafers, and chips are suitable to be used with Epo-Fix. Kit consists of: 1 kg resin, 120 g hardener, 15 cups, 15 stirrers, 1 x 5 mL and 20 mL syringes.