Learn More
STEMCELL Technologies EPO Immunoaffinity Isolation Plate, Size: 1 Unit
SDP
Catalog No. 501974248
Encompass
EPO Immunoaffinity Isolation Plate is a 96-well microtiter plate coated with a high-affinity monoclonal antibody (anti-EPO-26) against human erythropoietin (EPO). Each well can be used for immunoaffinity purification of natural or recombinant human EPO from serum, plasma, urine or other samples. EPO isolated using the EPO Immunaffinity Isolation Plate is appropriate for further analysis by methods such as SDS-PAGE, isoelectric focusing and mass spectrometry.
Catalog No. 50-197-4248 Supplier STEMCELL Technologies Supplier No. 01640
EPO Immunoaffinity Isolation Plate is a 96-well microtiter plate coated with a high-affinity monoclonal antibody (anti-EPO-26) against human erythropoietin (EPO). Each well can be used for immunoaffinity purification of natural or recombinant human EPO from serum, plasma, urine or other samples. EPO isolated using the EPO Immunaffinity Isolation Plate is appropriate for further analysis by methods such as SDS-PAGE, isoelectric focusing and mass spectrometry.