Learn More
Cayman Chemical EPO mRNACap-1UTP 100ug
SDP
Catalog No. 502945515
Encompass_Preferred
An mRNA for the expression of EPO; encapsulation in LNPs can be used for mRNA delivery and expression of EPO in vitro or in vivo
Catalog No. 50-294-5515 Supplier Cayman Chemical Supplier No. 41961100 UG
An mRNA for the expression of EPO; encapsulation in LNPs can be used for mRNA delivery and expression of EPO in vitro or in vivo