Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10075MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Mammal. Species Reactivity: Macaque. Protein Expression: A DNA sequence encoding the Macaca mulatta (Rhesus macaque) Erythropoietin / EPO, was expressed in the hosts and tags indicated. AA Sequence: APPRLVCDSR VLERYLLEAK EAENVTMGCS ESCSLNENIT VPDTKVNFYA WKRIEVGQQA VEVWQGLALL SEAVLRGQAV LANSSQPFEP LQLHMDKAIS GLRSITTLLR ALGAQEAISL PDAASAAPLR TITADTFCKL FRVYSNFLRG KLKLYTGEAC RRGDR. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: EPO. Gene ID: 719294. Uniprot: Q28513.
Catalog No. 50-188-5487