Learn More
Genscript Corporation EPO R/Erythropoietin R hFc Chi
Catalog No. 5000059518
Encompass
The erythropoietin (epo) receptor is a member of the cytokine receptor family It is expressed almost exclusively on erythroid precursor cells and controls the development of red blood cells The epo receptor has no intrinsic kinase activity but binds intracellular tyrosine kinases to elicit its signals Alterations in the transmission of the signalling cascade lead to clinically abnormal red blood cell production
Catalog No. 50-000-59518 Supplier Genscript Corporation Supplier No. Z04447500
The erythropoietin (epo) receptor is a member of the cytokine receptor family It is expressed almost exclusively on erythroid precursor cells and controls the development of red blood cells The epo receptor has no intrinsic kinase activity but binds intracellular tyrosine kinases to elicit its signals Alterations in the transmission of the signalling cascade lead to clinically abnormal red blood cell production