Learn More
OriGene EPO Rabbit Polyclonal Antibody, 100 uL
SDP
Supplier:  OriGene TA351162
Encompass_Preferred
This gene is a member of the EPO/TPO family and encodes a secreted, glycosylated cytokine composed of four alpha helical bundles. The protein is found in the plasma and regulates red cell production by promoting erythroid differentiation and initiating hemoglobin synthesis. This protein also has neuroprotective activity against a variety of potential brain injuries and antiapoptotic functions in several tissue types.
Catalog No. NC1850623