Learn More
Electron Microscopy Sciences Epo-Tek H20E  Adhesive 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 1267120E
Encompass_Preferred
Epo-Tek H20E is a two component, 100% solid silver-filled epoxy system, silver-resin paste and liquid hardener, mixing ration is 1:1. Epo-Tek H22E  features high thermal conductivity, and is very well suited for extensive high temperature applications (300 – 400 degree C)
Catalog No. 50-338-42