Learn More
Electron Microscopy Sciences Epo-Tek H20S  Adhesive 1 OZ
SDP
Catalog No. 5033843
Encompass_Preferred
Epo-Tek H20S is a modified version of Epo-TekH20E. Epo-Tek H20S is a highly reliable, two component, silver-filled epoxy with a smooth, thixotropic consistency (mixing ratio 1:1). This modified version offers high electrical conductivity, short curing cycles, proven reliability, and the convenient mix ratio, Epo-Tek H20S is extremely simple to use. Epo-Tek H20S pot life is 2.5 days and shelf life is one year when store at room temperature.
Catalog No. 50-338-43 Supplier Electron Microscopy Sciences Supplier No. 1267220S
Epo-Tek H20S is a modified version of Epo-TekH20E. Epo-Tek H20S is a highly reliable, two component, silver-filled epoxy with a smooth, thixotropic consistency (mixing ratio 1:1). This modified version offers high electrical conductivity, short curing cycles, proven reliability, and the convenient mix ratio, Epo-Tek H20S is extremely simple to use. Epo-Tek H20S pot life is 2.5 days and shelf life is one year when store at room temperature.