Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Epoc™ Host2 Motorola

Catalog No. 22347439 Shop All Siemens Healthineers Products
Change view
Click to view available options
For Use With (Equipment):
Epoc™ Reader
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-347-439 Epoc™ Reader
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-347-439 Supplier Siemens Healthineers Supplier No. 10736387

missing translation for 'validMainframeMsg'

Siemens Healthineers Epoc™ Host2 Motorola is a wireless handheld solution that provides blood gas, electrolyte, and metabolite results at the patient's side in approximately 35 seconds.

  • Reduces risk with patient-side testing that allows positive patient ID and eliminates transcription errors
  • Easily and securely connects to the Siemens Healthineers open POC informatics solution POCcelerator™ Software
  • Enables providers or healthcare systems to chose reference ranges to suit their clinical pathways
  • Accurate results are immediately trasmitted into the EMR
  • Compatible with Microsoft Windows Mobile 6.5 Classic
  • Use of a single cartridge reduces cost and improves workflow
  • Contains an expansive test menu with 13 different analytes
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Epoc™ Reader
Dimensions (L x W) 5.78 x 3.03 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.