Learn More
Eurofins Discoverx PathHunter Epoetin Alfa Bioassay Kit, 2-Plate. (93-0965Y3-00017)
Catalog No. 502001696
Encompass
The PathHunter Epoetin Alfa Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses erythropoetin or epoetin alfa-induced dimerization of EPOR receptors. This product uses EFC technology. The cell line included overexpresses EPOR target protein fused to PK alongside EPOR target protein fused to EA. Upon ligand-induced dimerization of EPOR, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1696 Supplier Eurofins Discoverx Supplier No. 930965Y300017
The PathHunter Epoetin Alfa Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses erythropoetin or epoetin alfa-induced dimerization of EPOR receptors. This product uses EFC technology. The cell line included overexpresses EPOR target protein fused to PK alongside EPOR target protein fused to EA. Upon ligand-induced dimerization of EPOR, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.